2-[4-(Aminocarbonyl)phenoxy]-4-methyl-1,3-thiazole-5-carboxylic acid

Supplier: Matrix Scientific

077098-500MG 077098-1G 077098-5G
102664-452EA 387.69 USD
102664-452 102664-454 102664-456
2-[4-(Aminocarbonyl)phenoxy]-4-methyl-1,3-thiazole-5-carboxylic acid
2-[4-(Aminocarbonyl)phenoxy]-4-methyl-1,3-thiazole-5-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀N₂O₄S
MDL Number: MFCD15731796
CAS Number: 1228551-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR