N-(4-Methoxyphenyl)-2-piperazin-1-ylacetamide dihydrochloride

Supplier: Matrix Scientific

077090-500MG 077090-1G
102664-424EA 516.92 USD
102664-424 102664-426
N-(4-Methoxyphenyl)-2-piperazin-1-ylacetamide dihydrochloride
N-(4-Methoxyphenyl)-2-piperazin-1-ylacetamide dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₁Cl₂N₃O₂
MDL Number: MFCD13857446
CAS Number: 1186049-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR