4-Amino-5-(3-thienyl)-4H-1,2,4-triazole-3-thiol

Supplier: Matrix Scientific

077088-500MG 077088-1G
102664-414EA 516.92 USD
102664-414 102664-416
4-Amino-5-(3-thienyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(3-thienyl)-4H-1,2,4-triazole-3-thiol

Hazard: Irritant

Formula: C₆H₆N₄S₂
MDL Number: MFCD10690331
CAS Number: 1092308-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR