tert-Butyl-4-formyl-2,6-dimethoxyphenyl carbonate ≥95%

Supplier: Matrix Scientific

077051-500MG 077051-1G
102664-326EA 298.46 USD
102664-326 102664-328
tert-Butyl-4-formyl-2,6-dimethoxyphenyl carbonate ≥95%
tert-Butyl-4-formyl-2,6-dimethoxyphenyl carbonate

Hazard: Irritant

Formula: C₁₄H₁₈O₆
MW: 282.3 g/mol
MDL Number: MFCD22383950
CAS Number: 1393845-71-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR