Methyl-6-{bis[(tert-butoxy)carbonyl]amino}-5-bromopyridine-3-carboxylate ≥97%
Supplier: Matrix Scientific
077041-500MG
077041-1G
077041-5G
102664-294EA
298.46
USD
102664-294
102664-296
102664-298
Methyl-6-{bis[(tert-butoxy)carbonyl]amino}-5-bromopyridine-3-carboxylate ≥97%
Methyl-6-{bis[(tert-butoxy)carbonyl]amino}-5-bromopyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₃BrN₂O₆ MW: 431.29 g/mol Melting Pt: 129…131 °C |
MDL Number:
MFCD22383953 CAS Number: 1393845-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...