N-(3-Iodoquinolin-8-yl)acetamide ≥97%

Supplier: Matrix Scientific

077038-500MG 077038-1G
102664-282EA 600 USD
102664-282 102664-284
N-(3-Iodoquinolin-8-yl)acetamide ≥97%
N-(3-Iodoquinolin-8-yl)acetamide

Hazard: Irritant

Formula: C₁₁H₉IN₂O
Melting Pt: 127…135 °C
MDL Number: MFCD22383944
CAS Number: 1393845-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR