4-Iodo-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

077036-500MG 077036-1G
102664-274EA 600 USD
102664-274 102664-276
4-Iodo-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
4-Iodo-1-methyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₆INO₂
Melting Pt: 141…144 °C
MDL Number: MFCD22383938
CAS Number: 1393845-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR