Ethyl-2-[(4-cyanophenyl)amino]pyrimidine-4-carboxylate ≥95%
Supplier: Matrix Scientific
077030-500MG
077030-1G
102664-248EA
995.38
USD
102664-248
102664-250
Ethyl-2-[(4-cyanophenyl)amino]pyrimidine-4-carboxylate ≥95%
Ethyl-2-[(4-cyanophenyl)amino]pyrimidine-4-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₂N₄O₂ MW: 268.28 g/mol Melting Pt: 176…178 °C |
MDL Number:
MFCD22383954 CAS Number: 1393845-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...