Ethyl-N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]carbamothioyl}carbamate ≥95%

Supplier: Matrix Scientific

077029-500MG 077029-1G
102664-244EA 193.85 USD
102664-244 102664-246
Ethyl-N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]carbamothioyl}carbamate ≥95%
Ethyl-N-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]carbamothioyl}carbamate

Hazard: Irritant

Formula: C₁₀H₉ClF₃N₃O₂S
MW: 327.71 g/mol
MDL Number: MFCD22383949
CAS Number: 1206641-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR