1-(2,3-Dihydro-1H-inden-2-yl)-2-methylpropan-2-amine hydrochloride ≥95%
Supplier: Matrix Scientific
077020-500MG
077020-1G
102664-210EA
600
USD
102664-210
102664-212
1-(2,3-Dihydro-1H-inden-2-yl)-2-methylpropan-2-amine hydrochloride ≥95%
1-(2,3-Dihydro-1H-inden-2-yl)-2-methylpropan-2-amine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀ClN |
MDL Number:
MFCD22383947 CAS Number: 1034457-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...