3-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}propane-1,2-diol ≥95%
Supplier: Matrix Scientific
077014-500MG
077014-1G
102664-184EA
298.46
USD
102664-184
102664-186
3-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}propane-1,2-diol ≥95%
3-{[3-Chloro-5-(trifluoromethyl)pyridin-2-yl]amino}propane-1,2-diol
Hazard: Irritant
Formula:
C₉H₁₀ClF₃N₂O₂ |
MDL Number:
MFCD22383956 CAS Number: 1393845-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...