2-Bromo-1-(diethoxymethyl)-4-fluorobenzene ≥97%

Supplier: Matrix Scientific

077006-500MG 077006-1G
102664-154EA 298.46 USD
102664-154 102664-156
2-Bromo-1-(diethoxymethyl)-4-fluorobenzene ≥97%
2-Bromo-1-(diethoxymethyl)-4-fluorobenzene

Hazard: Irritant

Formula: C₁₁H₁₄BrFO₂
MDL Number: MFCD22383957
CAS Number: 198967-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR