tert-Butyl-4-[(2-fluoro-5-nitrophenyl)carbonyl]piperazine-1-carboxylate ≥97%

Supplier: Matrix Scientific

076663-500MG 076663-1G
102547-330EA 298.46 USD
102547-330 102547-332
tert-Butyl-4-[(2-fluoro-5-nitrophenyl)carbonyl]piperazine-1-carboxylate ≥97%
tert-Butyl-4-[(2-fluoro-5-nitrophenyl)carbonyl]piperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₆H₂₀FN₃O₅
MW: 353.35 g/mol
Melting Pt: 117…118 °C
MDL Number: MFCD22373671
CAS Number: 1379526-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR