tert-Butyl-4-[(2-amino-3-methylphenyl)carbonyl]piperazine-1-carboxylate ≥97%
Supplier: Matrix Scientific
076662-500MG
076662-1G
076662-5G
102547-324EA
600
USD
102547-324
102547-326
102547-328
tert-Butyl-4-[(2-amino-3-methylphenyl)carbonyl]piperazine-1-carboxylate ≥97%
tert-Butyl-4-[(2-amino-3-methylphenyl)carbonyl]piperazine-1-carboxylate
Hazard: Irritant
Formula:
C₁₇H₂₅N₃O₃ Melting Pt: 91…92 °C |
MDL Number:
MFCD22373670 CAS Number: 1379527-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...