4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid ≥97%

Supplier: Matrix Scientific

076653-500MG 076653-1G
102547-288EA 600 USD
102547-288 102547-290
4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid ≥97%
4-Methyl-2-[4-(trifluoromethyl)phenyl]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₁F₃O₂
MDL Number: MFCD18425410
CAS Number: 273727-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR