Methyl-2-methyl-2-[(4-methylphenyl)amino]propanoate ≥95%
Supplier: Matrix Scientific
076647-500MG
076647-1G
076647-5G
102547-262EA
600
USD
102547-262
102547-264
102547-266
Methyl-2-methyl-2-[(4-methylphenyl)amino]propanoate ≥95%
Methyl-2-methyl-2-[(4-methylphenyl)amino]propanoate
Hazard: Irritant
Formula:
C₁₂H₁₇NO₂ MW: 207.27 g/mol |
MDL Number:
MFCD12817867 CAS Number: 1183683-27-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...