4-{2-Methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine dihydrochloride ≥97%

Supplier: Matrix Scientific

076646-500MG 076646-1G
102547-258EA 543.08 USD
102547-258 102547-260
4-{2-Methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine dihydrochloride ≥97%
4-{2-Methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈Cl₂N₄
MW: 289.21 g/mol
Melting Pt: 190…195 °C
MDL Number: MFCD22373666
CAS Number: 1379527-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR