Methyl-2-{[(4-chlorophenyl)methyl]amino}-2-methylpropanoate ≥95%

Supplier: Matrix Scientific

076643-500MG 076643-1G
102547-246EA 600 USD
102547-246 102547-248
Methyl-2-{[(4-chlorophenyl)methyl]amino}-2-methylpropanoate ≥95%
Methyl-2-{[(4-chlorophenyl)methyl]amino}-2-methylpropanoate

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₂
MDL Number: MFCD12039971
CAS Number: 1157807-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR