Ethyl-6-{2,4-dioxo-3-azatricyclo[7.3.1.0

Supplier: Matrix Scientific

Synonyms: Ethyl 6-(1,3-dioxo-1H-benzo[de]isoquinolin-2(3H)-yl)hexanoate

076633-500MG 076633-1G
102547-198EA 403.08 USD
102547-198 102547-200
Ethyl-6-{2,4-dioxo-3-azatricyclo[7.3.1.0
Ethyl-6-{2,4-dioxo-3-azatricyclo[7.3.1.0^{5,13}]trideca-1(13),5,7,9,11- pentaen-3-yl}hexanoate

Hazard: Irritant

Formula: C₂₀H₂₁NO₄
MW: 339.39 g/mol
Melting Pt: 88…90 °C
MDL Number: MFCD00376486
CAS Number: 300815-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR