Ethyl-4-chloro-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate ≥97%

Supplier: Matrix Scientific

076630-500MG 076630-1G 076630-5G
102547-180EA 600 USD
102547-180 102547-182 102547-184
Ethyl-4-chloro-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate ≥97%
Ethyl-4-chloro-5-(trifluoromethyl)-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₇H₆ClF₃N₂O₂
MW: 242.59 g/mol
Melting Pt: 115…117 °C
MDL Number: MFCD22373662
CAS Number: 855345-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR