Ethyl-5-bromo-4-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate ≥97%

Supplier: Matrix Scientific

076629-500MG 076629-1G
102547-176EA 995.38 USD
102547-176 102547-178
Ethyl-5-bromo-4-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate ≥97%
Ethyl-5-bromo-4-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

Hazard: Irritant

Formula: C₉H₉BrClNO₃
MW: 294.54 g/mol
Melting Pt: 139…140 °C
MDL Number: MFCD22373661
CAS Number: 1379526-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR