6-[Cyano(phenyl)methyl]pyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

076623-500MG 076623-1G
102547-148EA 384.62 USD
102547-148 102547-150
6-[Cyano(phenyl)methyl]pyridine-2-carboxylic acid ≥95%
6-[Cyano(phenyl)methyl]pyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₀N₂O₂
Melting Pt: 150 °C (dec.)
MDL Number: MFCD22373659
CAS Number: 1379527-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR