1-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidin-2-one ≥95%
Supplier: Matrix Scientific
076621-500MG
076621-1G
102547-140EA
673.85
USD
102547-140
102547-142
1-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidin-2-one ≥95%
1-[(4-Chlorophenyl)methyl]-5,5-dimethylimidazolidin-2-one
Hazard: Irritant
Formula:
C₁₂H₁₅ClN₂O Melting Pt: 122…126 °C |
MDL Number:
MFCD22373658 CAS Number: 1379527-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...