5-(4-Chlorophenyl)-3-iodopyridin-2-amine ≥97%

Supplier: Matrix Scientific

076620-500MG 076620-1G
102547-136EA 600 USD
102547-136 102547-138
5-(4-Chlorophenyl)-3-iodopyridin-2-amine ≥97%
5-(4-Chlorophenyl)-3-iodopyridin-2-amine

Hazard: Irritant

Formula: C₁₁H₈ClIN₂
Melting Pt: 136…137 °C
MDL Number: MFCD22373657
CAS Number: 1356397-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR