5,7-Bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

076614-500MG 076614-1G 076614-5G
102547-102EA 889.23 USD
102547-102 102547-104 102547-106
5,7-Bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid ≥95%
5,7-Bis(trifluoromethyl)-1,8-naphthyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₄F₆N₂O₂
Melting Pt: 171 °C (dec.)
MDL Number: MFCD22373654
CAS Number: 1379526-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR