(1-Azaniumyl-2-methylpropan-2-yl)(benzyl)azanium dichloride ≥95%

Supplier: Matrix Scientific

Synonyms: N-Benzyl-2-methylpropane-1,2-diamine dihydrochloride

076612-500MG 076612-1G
102547-092EA 803.08 USD
102547-092 102547-094
(1-Azaniumyl-2-methylpropan-2-yl)(benzyl)azanium dichloride ≥95%
(1-Azaniumyl-2-methylpropan-2-yl)(benzyl)azanium dichloride

Hazard: Irritant

Formula: C₁₁H₂₀Cl₂N₂
MW: 251.2 g/mol
Melting Pt: 168…172 °C
MDL Number: MFCD22373653
CAS Number: 1379527-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR