H-Phe-Phe-OH . HCl ≥95%

Supplier: Matrix Scientific

073331-1G 073331-5G 073331-10G
102529-602EA 213.85 USD
102529-602 102529-604 102529-606
H-Phe-Phe-OH . HCl ≥95%
H-Phe-Phe-OH . HCl

Hazard: Irritant

Formula: C₁₈H₂₁ClN₂O₃
MDL Number: MFCD22631441
CAS Number: 68800-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR