6-(Trifluoromethyl)pyrimidin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Amino-6-(trifluoromethyl)pyrimidine

072606-250MG 072606-1G
102525-764EA 676.92 USD
102525-764 102525-766
6-(Trifluoromethyl)pyrimidin-4-amine ≥95%
6-(Trifluoromethyl)pyrimidin-4-amine

Hazard: Irritant

Formula: C5H4F3N3
MW: 163.1000 g/mol
MDL Number: MFCD08436657
CAS Number: 672-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR