1-(4-Chloro-3-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroquinoline ≥95%
Supplier: Matrix Scientific
071961-1G
071961-5G
071961-10G
102522-330EA
249.23
USD
102522-330
102522-332
102522-334
1-(4-Chloro-3-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroquinoline ≥95%
1-(4-Chloro-3-nitrobenzenesulfonyl)-1,2,3,4-tetrahydroquinoline
Hazard: Irritant
Formula:
C₁₅H₁₃ClN₂O₄S MW: 352.7900 g/mol |
MDL Number:
MFCD08275686 CAS Number: 847170-51-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...