tert-Butyl-4-{2-methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate ≥97%

Supplier: Matrix Scientific

071448-500MG 071448-1G
102520-104EA 600 USD
102520-104 102520-106
tert-Butyl-4-{2-methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate ≥97%
tert-Butyl-4-{2-methyl-1H-imidazo[4,5-b]pyridin-1-yl}piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₄N₄O₂
MW: 316.41 g/mol
Melting Pt: 197…200 °C
MDL Number: MFCD22126124
CAS Number: 1373350-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR