2-Methyl-2-{[(4-methylphenyl)methyl]amino}propanenitrile ≥95%

Supplier: Matrix Scientific

071444-500MG 071444-1G
102520-092EA 298.46 USD
102520-092 102520-094
2-Methyl-2-{[(4-methylphenyl)methyl]amino}propanenitrile ≥95%
2-Methyl-2-{[(4-methylphenyl)methyl]amino}propanenitrile

Hazard: Irritant

Formula: C₁₂H₁₆N₂
Melting Pt: 36…40 °C
MDL Number: MFCD10014475
CAS Number: 1018265-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR