3-Iodo-8-nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

071439-500MG 071439-1G
102520-072EA 403.08 USD
102520-072 102520-074
3-Iodo-8-nitroimidazo[1,2-a]pyridine ≥97%
3-Iodo-8-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₇H₄IN₃O₂
Melting Pt: 222…224 °C
MDL Number: MFCD22126118
CAS Number: 1373350-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR