Ethyl-2-nitro-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoate ≥97%
Supplier: Matrix Scientific
071435-500MG
071435-1G
102520-058EA
298.46
USD
102520-058
102520-060
Ethyl-2-nitro-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoate ≥97%
Ethyl-2-nitro-3-{1H-pyrrolo[2,3-b]pyridin-3-yl}propanoate
Hazard: Irritant
Formula:
C₁₂H₁₃N₃O₄ MW: 263.26 g/mol Melting Pt: 136…137 °C |
MDL Number:
MFCD22126111 CAS Number: 1373348-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...