5-(1,3-Dioxan-2-yl)-1-methyl-1H-indazol-3-amine ≥95%

Supplier: Matrix Scientific

071434-500MG 071434-1G
102520-054EA 403.08 USD
102520-054 102520-056
5-(1,3-Dioxan-2-yl)-1-methyl-1H-indazol-3-amine ≥95%
5-(1,3-Dioxan-2-yl)-1-methyl-1H-indazol-3-amine

Hazard: Irritant

Formula: C₁₂H₁₅N₃O₂
Melting Pt: 191…193 °C
MDL Number: MFCD22126123
CAS Number: 1373350-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR