1-Cyclohexyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one ≥95%

Supplier: Matrix Scientific

071430-500MG 071430-1G
102520-038EA 298.46 USD
102520-038 102520-040
1-Cyclohexyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one ≥95%
1-Cyclohexyl-1H,2H,3H-imidazo[4,5-b]pyridin-2-one

Hazard: Irritant

Formula: C₁₂H₁₅N₃O
Melting Pt: 224…228 °C
MDL Number: MFCD22126108
CAS Number: 906532-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR