4-[(6-Chloropyridin-3-yl)methyl]benzaldehyde ≥95%
Supplier: Matrix Scientific
071428-500MG
071428-1G
102520-030EA
298.46
USD
102520-030
102520-032
4-[(6-Chloropyridin-3-yl)methyl]benzaldehyde ≥95%
4-[(6-Chloropyridin-3-yl)methyl]benzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₀ClNO Melting Pt: 90…92 °C |
MDL Number:
MFCD22126117 CAS Number: 1373348-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...