3-Bromo-1H-indole-2-carbonitrile ≥95%

Supplier: Matrix Scientific

071421-500MG 071421-1G
102520-004EA 803.08 USD
102520-004 102520-006
3-Bromo-1H-indole-2-carbonitrile ≥95%
3-Bromo-1H-indole-2-carbonitrile

Hazard: Irritant

Formula: C₉H₅BrN₂
Melting Pt: 176…178 °C
MDL Number: MFCD22126115
CAS Number: 1373348-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR