1-Benzyl-5,5-dimethylimidazolidin-2-one ≥97%

Supplier: Matrix Scientific

071419-500MG 071419-1G
102519-996EA 384.62 USD
102519-996 102519-998
1-Benzyl-5,5-dimethylimidazolidin-2-one ≥97%
1-Benzyl-5,5-dimethylimidazolidin-2-one

Hazard: Irritant

Formula: C₁₂H₁₆N₂O
Melting Pt: 142…146 °C
MDL Number: MFCD22126126
CAS Number: 1373350-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR