5-Amino-1-benzyl-1H-pyrrole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

071416-500MG 071416-1G
102519-986EA 261.54 USD
102519-986 102519-988
5-Amino-1-benzyl-1H-pyrrole-3-carbonitrile ≥95%
5-Amino-1-benzyl-1H-pyrrole-3-carbonitrile

Hazard: Irritant

Formula: C₁₂H₁₁N₃
Melting Pt: 54…56 °C
MDL Number: MFCD20700824
CAS Number: 59661-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR