2-Methyl-2-[(2-methylpropyl)amino]propanenitrile ≥95%

Supplier: Matrix Scientific

071363-500MG 071363-1G
102519-898EA 403.08 USD
102519-898 102519-900
2-Methyl-2-[(2-methylpropyl)amino]propanenitrile ≥95%
2-Methyl-2-[(2-methylpropyl)amino]propanenitrile

Hazard: Irritant

Formula: C₈H₁₆N₂
MDL Number: MFCD10014464
CAS Number: 1018291-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR