Methyl-3-cyano-1-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate ≥97%

Supplier: Matrix Scientific

071357-500MG 071357-1G
102519-874EA 600 USD
102519-874 102519-876
Methyl-3-cyano-1-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate ≥97%
Methyl-3-cyano-1-methyl-1H-pyrrolo[2,3-b]pyridine-5-carboxylate

Hazard: Irritant

Formula: C₁₁H₉N₃O₂
Melting Pt: 175…176 °C
MDL Number: MFCD22126104
CAS Number: 1370587-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR