Methyl-4-[(6-chloropyridin-3-yl)methyl]benzoate ≥97%

Supplier: Matrix Scientific

071356-500MG 071356-1G
102519-870EA 363.08 USD
102519-870 102519-872
Methyl-4-[(6-chloropyridin-3-yl)methyl]benzoate ≥97%
Methyl-4-[(6-chloropyridin-3-yl)methyl]benzoate

Hazard: Irritant

Formula: C₁₄H₁₂ClNO₂
Melting Pt: 80…83 °C
MDL Number: MFCD22126099
CAS Number: 1370587-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR