6-(4-Methoxyphenyl)pyridine-3-carbaldehyde ≥99%

Supplier: Matrix Scientific

071353-500MG 071353-1G 071353-5G
102519-856EA 403.08 USD
102519-856 102519-858 102519-860
6-(4-Methoxyphenyl)pyridine-3-carbaldehyde ≥99%
6-(4-Methoxyphenyl)pyridine-3-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
Melting Pt: 116…118 °C
MDL Number: MFCD04115433
CAS Number: 834884-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR