Ethyl-2-chloro-5-nitropyridine-4-carboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-chloro-5-nitroisonicotinate
071340-500MG
071340-1G
102519-802EA
607.69
USD
102519-802
102519-804
Ethyl-2-chloro-5-nitropyridine-4-carboxylate ≥97%
Ethyl-2-chloro-5-nitropyridine-4-carboxylate
Hazard: Irritant
Formula:
C₈H₇ClN₂O₄ MW: 230.61 g/mol Melting Pt: 40…41 °C |
MDL Number:
MFCD22126101 CAS Number: 907545-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...