2-Chloro-5-[(4-methoxyphenyl)methyl]pyridine ≥95%

Supplier: Matrix Scientific

071330-500MG 071330-1G 071330-5G
102519-760EA 298.46 USD
102519-760 102519-762 102519-764
2-Chloro-5-[(4-methoxyphenyl)methyl]pyridine ≥95%
2-Chloro-5-[(4-methoxyphenyl)methyl]pyridine

Hazard: Irritant

Formula: C₁₃H₁₂ClNO
Melting Pt: 46…49 °C
MDL Number: MFCD22126116
CAS Number: 1370587-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR