5-Benzyl-2-chloropyridine ≥97%

Supplier: Matrix Scientific

071322-500MG 071322-1G
102519-734EA 307.69 USD
102519-734 102519-736
5-Benzyl-2-chloropyridine ≥97%
5-Benzyl-2-chloropyridine

Hazard: Irritant

Formula: C₁₂H₁₀ClN
Melting Pt: 48…50 °C
MDL Number: MFCD22126098
CAS Number: 149879-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR