Ethyl-2-chloro-4-methoxypyrimidine-5-carboxylate ≥97%

Supplier: Matrix Scientific

071104-5G 071104-1G
102519-476EA 2462.15 USD
102519-476 102833-488
Ethyl-2-chloro-4-methoxypyrimidine-5-carboxylate ≥97%
Ethyl-2-chloro-4-methoxypyrimidine-5-carboxylate

Hazard: Irritant

Formula: C₈H₉ClN₂O₃
MDL Number: MFCD22199254
CAS Number: 1260178-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR