Methyl-4,6-dibromonicotinate ≥95%

Supplier: Matrix Scientific

071083-1G 071083-5G
102663-858EA 504.62 USD
102663-858 102663-860
Methyl-4,6-dibromonicotinate ≥95%
Methyl-4,6-dibromonicotinate

Hazard: Irritant

Formula: C₇H₅Br₂NO₂
MDL Number: MFCD00234262
CAS Number: 1364663-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR