Samarium ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGSM1-500ML
CGSM10-500ML
100360-694EA
133.54
USD
100360-694
100359-792
Samarium ICP Standard, Inorganic Ventures
Standards
Cationic Standards
Samarium Standards
Certified reference material for Samarium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).