Phosphorus ICP Standard, Inorganic Ventures
Supplier: INORGANIC VENTURES MS
CGP10-500ML
CGP1-500ML
100360-070EA
217.54
USD
100360-070
100360-630
Phosphorus ICP Standard, Inorganic Ventures
Standards
Cationic Standards
Phosphorus Standards
Certified reference material for Phosphorus, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).